Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotix Pure uTIP™ Sterile Pipette Tips
GREENER_CHOICE

Catalog No. p-200009117 Shop All Biotix Products
Click to view available options
Volume (Metric):
10 μL
200 μL
250 μL
300 μL
1000 μL
1250 μL
Tip Style:
Ergonomic, Extended Length, Fine Point, Fine Point, Finely Tapered, Flextop
Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
This item is not returnable. View return policy
This item is not returnable. View return policy

Pure uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard are designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Manufactured with Pure resin made of virgin polypropylene.

  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard
  • Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal.
  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin made of virgin polypropylene.
  • Fits all standard pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Compatibility Universal
Format Hinged Rack Sterile
Tip Style Ergonomic, Extended Length, Fine Point, Fine Point, Finely Tapered, Flextop
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit, Universal Fit Multichannel, Universal Fit Ultramicro
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less

This product contains no safety hazards

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.