Learn More
Gentex™ PureFlo 3000 Respirator, Open Frame
Click to view available options
:
Color: Gray
Description
- PF3000 Respirator
- Open Frame
- Clear Visor
- Tychem QC Face Seal
- No battery charger
Specifications
Specifications
Assigned Protection Factor (APF) | 25 |
Cartridge/Filter | PARTICULATE |
Color | Gray |
Product Type | PAPR Assembly, No Headtop |
Size | One Size |
Filter Efficiency (NIOSH) | HEPA |
Intrinsic Safety (IS) Rating | No |
IP Rating | IP54 |