Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo PRO2475 Fire Resistant Neck Cape
SDP

Catalog No. p-7225769
Click to view available options
Model:
PR02475
PR02475SP
Quantity:
1 Ea.
2/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Spare neck cape assembly for use with the PureFlo ESM+ PAPR.

  • Flame Resistant as per ASTM D6413
  • Abrasion Resistant
  • Flash Fire Protection as per ASTM F1930
  • Chemical Penetration Resistance as per ASTM F903
  • Adjustable neck cord
  • Two velcro attachments for securing free end of lanyard
  • Twelve snap fasteners for easy removal and attachment
  • Used with PureFlo ESM, ESM+ and Airline SAR

Specifications

Color Black
Disposable Reusable
Material Stedprene
Product Type Neck Cape
Size One Size
For Use With (Equipment) PureFlo ESM+, ESM, and PR02000 PAPR Helmet Systems
Includes Adjustable neck cord
Model PR02475
Quantity 1 Ea.
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.