Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo White Integrity 1800 Neck Cape
SDP

Catalog No. p-7225777
Click to view available options
Description:
Double Bib Neck Cape, 2/Pk.
Single Bib Neck Cape
Single Bib Neck Cape, 2/Pk.
Quantity:
1 Ea.
2/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Neck cape for use with the PureFlo ESM and ESM+ PAPR System. Suitable for Class 10 cleanroom environments.

  • White Integrity 1800 Material
  • Densely woven continuous filament polyester fabric with an ESD stripe
  • Designed to meet the quality and performance standards in critical environments
  • Highly fluid repellent (Teflon HT treatment)
  • Durable antimicrobial treatment (Aegis) to inhibit bacterial growth and reduce bioburden
  • Recommended for pharmaceutical, biotech and similar applications
  • Class 10 (ISO Class 4) and higher
  • Adjustable neck cord
  • Two Velcro attachments for securing free end of lanyard
  • Twelve snap fasteners for easy removal and attachment
  • Available in single or double bib options

Specifications

Description Single Bib Neck Cape
Color White
Disposable Single-use
Material Integrity 1800
Product Type Neck Cape
Size One Size
For Use With (Application) Class 10 (ISO 4) Cleanroom Environments, Pharmaceutical, Biotech
For Use With (Equipment) PureFlo ESM and ESM+ PAPR System
Model PR01470
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.