Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Visor Assembly
SDP

Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.

$235.00 - $1045.00

Specifications

Description Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.
Color Clear
Disposable Reusable
Product Type Visor
Size One Size
View More Specs

Products 2
Catalog Number Mfr. No. Model Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Model Quantity Price Quantity & Availability  
18-444-054
SDP
View Documents
Gentex™
PR02480SP
PR02480SP 1 Ea.
Each for $235.00
Only null left
Add to Cart
 
18-444-055
SDP
View Documents
Gentex™
PR02050SP
PR02050SP 4/Pk.
Pack of 4 for $1,045.00
Only null left
Add to Cart
 
Description

Description

  • Spare visor assembly (Non Welding Unit)
  • For use with the PureFlo ESM+ PAPR
  • Color: Clear
Specifications

Specifications

Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.
Reusable
One Size
Yes
Clear
Visor
PureFlo PF60 ESM+
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.