Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ DNase Set

Catalog No. 12185010
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-185-010 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-185-010 Supplier Invitrogen™ Supplier No. 12185010
Only null left
Add to Cart
Edge
Add to Cart

Includes

PureLink™ DNase (1500U, lyophilized), 10X DNase I Reaction Buffer (1mL), RNase-Free Water (1mL)

The RNase-free DNase set comes lyophilized and is optimized for on-column digestion of DNA using PureLink™ protocols.

Using the PureLink™ DNase Set
The RNase-free DNase set comes lyophilized and is optimized for on-column digestion of DNA using PureLink™ protocols. The degree of DNA digestion performed by PureLink™ DNase is optional for most PureLink™ RNA kits, due to the efficient removal of the majority of DNA by the PureLink™ technology; however, some downstream applications that are sensitive to small amounts of DNA may require further removal of DNA.

  • RNase-free for use in RNA purification
  • Optimized for on-column digestion
  • Lyophilized for stability

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage Contains:
• PureLink™ DNase (1500 U, lyophilized)
• 10X DNase I Reaction Buffer (1 mL)
• RNase-Free Water (1 mL)

Store at 4°C upon receipt. After reconstituted, PureLink™ DNase should be stored at -20°C.
No. of Reactions 50 Preps
Quantity 50 Preps
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.