Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Fast Low-Endotoxin Midi Plasmid Purification Kit

Catalog No. p-8492020
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
25 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A35892 25 Preps
A36227 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A35892 Supplier Invitrogen™ Supplier No. A35892
Only null left
Add to Cart
Edge
Add to Cart

The PureLink Fast Low-Endotoxin Midi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (<1 EU/μg) plasmid DNA in about 30 minutes.

The simple precipitation-free protocol yields up to 0.4 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.

  • Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
  • Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
  • High yields—isolate up to 0.4 mg of high-quality plasmid DNA
  • Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Resuspension buffer, store at 4°C upon reciept
• Lysis buffer, store at room temperature
• Precipitation buffer, store at room temperature
• Binding buffer, store at room temperature
• Wash buffer 1, store at room temperature
• Wash buffer 2 (concentrate), store at room temperature
• Elution buffer, store at room temperature
• Column assemblies, store at room temperature
• Syringe filters, store at room temperature
• Syringe plungers, store at room temperature
• Collection tubes, store at room temperature

Format Kit
Isolation Technology Silica Column
Scale Midi
For Use With (Application) Cloning, PCR
No. of Reactions 25 Preps
Prep Scale Midi scale input culture of 25-100 mL to obtain yields of up to 200 μg
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 25 Preps
Shipping Condition Room Temperature
Test Time 30 min.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.