Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Pro 96 total RNA Purification Kit

Catalog No. p-200109802
Encompass_Preferred
Click to view available options
Quantity:
2 x 1 L
384 Preps
5 x 1 L
750 mL
Product Type:
RNA Lysis Buffer
RNA Purification Kit
RNA Wash Buffer I
RNA Wash Buffer II (5X)

The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes.

The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes. Isolate high yields of total RNA with low genomic DNA contamination from bacteria, yeast, plant, or mammalian samples.

The one-hour procedure requires no handling or disposal of organic waste. RNA binds to a silica membrane and impurities are removed by washing through vacuum filtration or centrifugation. High-purity total RNA is eluted into RNase-free water; ideal for use in a variety of applications, including RT-PCR, qPCR, northern blotting, and nuclease protection assays.

Improved plate design

The PureLink Pro 96 total RNA Purification Kit provides optimized semi-skirted 96-well plates(Figure 3) for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation (at 2250 x g), or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 150 mL PureLink Pro 96 Lysis Buffer; store at room temperature
• 250 mL PureLink Pro 96 Wash Buffer I; room temperature
• 250 mL PureLink Pro 96 Wash Buffer II; room temperature
• 75 mL PureLink Pro 96 RNase-free Water; room temperature
• 4 PureLink 96-well Total RNA Filter Plates; room temperature
• 4 PureLink Pro 96 Elution Plates; room temperature
Isolation Technology Silica Plate
Purification Time 25 min. (After sample homogenization)
Elution Volume 45–170 μL
Sample Type Bacteria, Cells, Plant, Tissue, Yeast
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) Biomek FX, Robotic Liquid Handling Systems, Eppendorf epMotion TECAN Freedom Evo, TECAN Genesis
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type RNA Purification Kit
Quantity 384 Preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤109 cells, Cells: varies, Plant: ≤150 mg, Tissue: ≤100 mg, Yeast: ≤1.8 mL
Yield Up to 33 μg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.