Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Soothing Gel
SDP

Catalog No. p-7159165
Click to view available options
Color:
Clear
Green
Packaging:
Portable Flip Cap Bottle
Portable Pump Bottle
Refill
Table Top Pump Bottle
Quantity:
12/Pk.
24/Pk.
8/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains aloe and vitamin E.

  • Kills most illness causing germs without damaging skin
  • Formulated with moisturizers and skin conditioners to keep hands soft
  • Refill pack option is SANITARY SEALED™ to prevent contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sterilizer/Disinfectant Hand Gel
Color Green
Form Gel
Packaging Portable Flip Cap Bottle
Quantity 24/Pk.
Size 4 oz.
Volume (English) 4 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.