Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PURG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599039
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599039 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599039 Supplier Invitrogen™ Supplier No. PA599039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PURG Polyclonal Antibody for Western Blot, ELISA

The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. PURG lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PURG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PURG
Gene Accession No. Q9UJV8
Gene Alias 4930486B15Rik; MGC119274; Purg; PURG-A; Pur-gamma; PURG-B; purine rich element binding protein G; purine-rich element binding protein G; Purine-rich element-binding protein G; Purine-rich element-binding protein gamma
Gene Symbols PURG
Host Species Rabbit
Immunogen Recombinant Human Purine-rich element-binding protein gamma protein (242-345AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29942
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.