Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Purified Mouse Anti-Human Neutrophil Elastase, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569518
Description
Neutrophil Elastase Monoclonal antibody specifically detects Neutrophil Elastase in Human samples. It is validated for Intracellular staining (flow cytometry)
Specifications
Neutrophil Elastase | |
Monoclonal | |
0.5 mg/mL | |
ELANE; HNE; PMN elastase; PMN-E; neutrophil elastase | |
Neutrophil granule proteins | |
0.1 mg | |
1991 | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
IgG1 |
Flow Cytometry | |
NP57 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction