Learn More
Specifications
Specifications
| Antigen | Purified mouse immunoglobin |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Host Species | Mouse |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Product Type | Antibody |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.