Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Purified Viral Lysate, Human Immunodeficiency Virus Type 2 (HIV-2)
Shop All ZeptoMetrix Corporation Products

Click to view available options
Quantity:
1 mg
Description
- Available in 1mg vials
Specifications
Specifications
Biosafety Level | 2 |
Certifications/Compliance | ISO, 13485 |
Concentration | 0.5 to 3mg/mL |
Content And Storage | -65°C |
Detects | Viral Antibodies |
Form | Solution |
Media Type | Vial |
Product Type | Purified Viral Lysate |
For Use With (Application) | Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay |
Purification Method | Sucrose density gradient ultracentrifugation |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction