Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purified Water, USP, Fisher Chemical™

Catalog No. w124 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
PolyPac™

Catalog No. W124

Only null left
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
CAS Min % 100%
Boiling Point 100°C
CAS Max % 100%
Description Filtered to 0.2 μm
Grade USP
Packaging Glass Bottle
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Purified Water
Show More Show Less
missing translation for 'healthHazard3'
Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
 
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use.
ShelfLife 5 Years
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.