Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier Axiom Class II Type C1 Biosafety Cabinet
SDP

Catalog No. 304411011 Shop All Labconco Corporation Products
Change view
Click to view available options
Width (English) Exterior:
54.3 in.
78.3 in.
Sash Opening (English):
8 in.
10 in.
Includes:
Accessory Pk.
Base Stand
Base Stand, Accessory Pk.
Biosafety Cabinet Only
Electrical Requirements:
115 V, 50/60 Hz, 12 A
230 V, 50/60 Hz, 6 A
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Exterior Sash Opening (English) Includes Electrical Requirements
30-441-1011 54.3 in. 10 in. Accessory Pk. 230 V, 50/60 Hz, 6 A
30-441-0001 54.3 in. 10 in. Biosafety Cabinet Only 115 V, 50/60 Hz, 12 A
30-441-0011 54.3 in. 10 in. Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-441-0101 54.3 in. 10 in. Base Stand 115 V, 50/60 Hz, 12 A
30-441-0111 54.3 in. 10 in. Base Stand 230 V, 50/60 Hz, 6 A
30-441-1001 54.3 in. 10 in. Accessory Pk. 115 V, 50/60 Hz, 12 A
30-441-1101 54.3 in. 10 in. Base Stand, Accessory Pk. 115 V, 50/60 Hz, 12 A
30-441-1111 54.3 in. 10 in. Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-448-0001 54.3 in. 8 in. Biosafety Cabinet Only 115 V, 50/60 Hz, 12 A
30-448-0011 54.3 in. 8 in. Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-448-0101 54.3 in. 8 in. Base Stand 115 V, 50/60 Hz, 12 A
30-448-0111 54.3 in. 8 in. Base Stand 230 V, 50/60 Hz, 6 A
30-448-1001 54.3 in. 8 in. Accessory Pk. 115 V, 50/60 Hz, 12 A
30-448-1011 54.3 in. 8 in. Accessory Pk. 230 V, 50/60 Hz, 6 A
30-448-1101 54.3 in. 8 in. Base Stand, Accessory Pk. 115 V, 50/60 Hz, 12 A
30-448-1111 54.3 in. 8 in. Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-461-0001 78.3 in. 10 in. Biosafety Cabinet Only 115 V, 50/60 Hz, 12 A
30-461-0011 78.3 in. 10 in. Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-461-0101 78.3 in. 10 in. Base Stand 115 V, 50/60 Hz, 12 A
30-461-0111 78.3 in. 10 in. Base Stand 230 V, 50/60 Hz, 6 A
30-461-1001 78.3 in. 10 in. Accessory Pk. 115 V, 50/60 Hz, 12 A
30-461-1011 78.3 in. 10 in. Accessory Pk. 230 V, 50/60 Hz, 6 A
30-461-1101 78.3 in. 10 in. Base Stand, Accessory Pk. 115 V, 50/60 Hz, 12 A
30-461-1111 78.3 in. 10 in. Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-0001 78.3 in. 8 in. Biosafety Cabinet Only 115 V, 50/60 Hz, 12 A
30-468-0011 78.3 in. 8 in. Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-468-0101 78.3 in. 8 in. Base Stand 115 V, 50/60 Hz, 12 A
30-468-0111 78.3 in. 8 in. Base Stand 230 V, 50/60 Hz, 6 A
30-468-1001 78.3 in. 8 in. Accessory Pk. 115 V, 50/60 Hz, 12 A
30-468-1011 78.3 in. 8 in. Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1101 78.3 in. 8 in. Base Stand, Accessory Pk. 115 V, 50/60 Hz, 12 A
30-468-1111 78.3 in. 8 in. Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 30-441-1011 Supplier Labconco™ Supplier No. 304411011
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Purifier Axiom Class II Type C1 Biosafety Cabinet is an unique, flexible BSC that can be operated as Type A or B and is the only Type C1 in the world.

  • Requires supporting base if included stand option is not selected
  • Intrinsically-safe negative pressure design includes:
    • Dual Electronically Commutated Motors (ECM)
    • Constant Airflow Profile™ (CAP) Technology airflow monitoring system
    • Active Protection Protocol operates the cabinet's exhaust blowers for up to five minutes preventing the flow of air into the laboratory in the event of exhaust system failure
    • BSC required Air-Wave™ Entry System at access opening to enhance containment
    • Contain-Air™ Negative Pressure Channel at top of sash to enhance containment
    • Two programmable working sash heights (8 in. and 10 in.)
    • 10° angled, fully-closing, 1/4 in. tempered safety glass sash with counterbalanced, anti-racking, easy-to-lift mechanism
    • Curved waterfall design stainless steel inlet grille with Reserve-Air™ Secondary Airflow Slots
    • 22.6 in. (58 cm) maximum sash opening height and 27.0 in. (69 cm) viewing height Chem-Zone™ work surface with dedicated direct exhaust for use with hazardous vapors or radionuclides
    • Leak-tight interior with removable, type 304 stainless steel 3-piece work surface (dished Chem-Zone in center and two flat sides) with lift out knobs and clearly delineated working area
    • Nominal inflow velocity of 105 feet per minute (fpm) (0.53 m/sec)
    • Nominal downflow velocity of 65 fpm (0.33 m/sec) for 4 ft. models and 55 fpm (0.28 m/sec) for 6 ft. models
  • Touchpad on right-hand side post controls blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection
  • Omni-Flex™ two mode design (Type A and B mode)
    • Low static pressure and volumetric rate exhaust requirements when in Type B mode
    • Night-Smart™ System for idling the blower when the sash is fully closed (while in Type A mode only)
  • MyLogic™ Operating System and interior-mounted, line-of-sight, full color, easy-to-understand LCD information center with:
    • Smart-Start™ System for start up and shut down operations
    • Filter Life Remaining bar graph for each HEPA filter
    • Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
    • Bright, 90-150 ft. candle, glare-free LED lighting located outside the contaminated work area
    • Interval/elapsed timer for experiment monitoring, LED light or UV light control (on models with UV light)
    • Electronic security lock (optional activation) that requires access code to operate the cabinet

Specifications

Description Class II Type C1 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Chamber Material Stainless Steel
Product Type Class II Biosafety Cabinet
Model Axiom
Noise Level 67 dBA
Outlets 2
Plug Type North America
Power Consumption 360 W
Receptacles 2
Sash Opening (Metric) 25 cm
Width (English) Exterior 54.3 in.
Volume (Metric) Exhaust 680 m3/hr.
Display Full-color LCD
Filter Efficiency 0.9999
Filter Type HEPA
Width (Metric) Exterior 137.9 cm
Housing Material Powder Coated Steel
Includes Accessory Pk.
Lighting LED
Sash Opening (English) 10 in.
Service Fixtures One
Warranty 5 Years
Height (English) Exterior 64.2 in.
Height (Metric) Exterior 163.1 cm
Depth (English) Exterior 32.7 in.
Depth (Metric) Exterior 83.1 cm
Net Weight (English) 636 lb.
Shipping Weight (Metric) 340.2 kg
Electrical Requirements 230 V, 50/60 Hz, 6 A
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.