Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Class I Safety Enclosure
SDP

Catalog No. 10304117 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Air Flow Monitor
No Plug
UV Lamp, Airflow Monitor
UV Lamp, w/o Airflow Monitor
Width (Metric):
60.3 cm
60.32 cm
90.8 cm
121.2 cm
121.28 cm
Electrical Requirements:
115 V, 60 Hz
230 V, 50 Hz
230 V, 50/60 Hz
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Width (Metric) Electrical Requirements
10-304-117 Air Flow Monitor 90.8 cm 230 V, 50 Hz
10-304-121 Air Flow Monitor 121.28 cm 230 V, 50/60 Hz
10-304-110 UV Lamp, w/o Airflow Monitor 60.32 cm 115 V, 60 Hz
10-304-112 No Plug 60.32 cm 230 V, 50/60 Hz
10-304-119 UV Lamp, Airflow Monitor 121.28 cm 115 V, 60 Hz
10-304-116 No Plug 90.8 cm 230 V, 50 Hz
10-304-113 No Plug 60.3 cm 230 V, 50 Hz
10-304-120 No Plug 121.2 cm 115 V, 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 10-304-117 Supplier Labconco™ Supplier No. 3980323
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Provide your staff with personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ Class I Safety Enclosure.

Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.

  • Total exhaust from 155 to 310cfm, depending on hood width
  • Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
  • Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
  • Airfoil with Clean-Sweep™ airflow openings
  • Optional built-in Guardian™ Airflow Monitor to alert user of face velocities outside preset range (where applicable)
  • With interlocked UV protection pane
  • Variable digital timer for UV lamp
  • Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
  • Class I Enclosures have a 15w, 254nm germicidal UV lamp to use in conjunction with surface disinfection to ensure decontamination of biohazardous particulates while unattended
  • 0.25 in. (6.4mm) Tempered glass side panels and sash
  • Glare-free fluorescent lamp mounted outside contamination area
  • Blower and light switches are front-mounted for easy access
  • Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
  • Back panel has two utility ports for service connections
  • Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface

For operator safety, UV lamp cannot be turned on when fluorescent lamp is in use.

Order Info

28.75L x 37 in. H (73 x 94cm). 115V 60Hz, 3A.

Certifications

ETL listed (certified to UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1).

Compliance

ASHRAE™ 110-1995 conformity tested. Biological containment verified per NSF Standard Number 49 Personnel Protection Test.

Specifications

Product Type Class I Safety Enclosure
Power Consumption 102 W
Height (English) 37.08 in.
Sash Opening (Metric) 23.9 cm
Includes Air Flow Monitor
Lighting Fluorescent
Height (Metric) 94.1 cm
Sash Opening (English) 12 in.
Width (English) 35.75 in.
Width (Metric) 90.8 cm
Frequency 50 Hz
Depth (English) 28.64 in.
Depth (Metric) 72.74 cm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Amperage 2 A
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.