Learn More
Labconco™ Purifier™ HEPA-Filtered Safety Enclosure with Guardian Monitor
Shop All Labconco Corporation Products

Description
Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.
- Total exhaust from 155 to 310cfm, depending on hood width
- Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
- Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
- Airfoil with Clean-Sweep™ airflow openings
- Built-in Guardian™ Airflow Monitor to alert user of face velocities outside preset range (where applicable)
- Front-mounted pressure gauge monitors pressure differential against the HEPA filter
- HEPA filter is 99.99% efficient on 0.3μm particles (where applicable)
- Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
- Class I Enclosures have a 15 W, 254 nm germicidal UV lamp to use in conjunction with surface disinfection to ensure decontamination of biohazardous particulates while unattended
- 0.25 in. (6.4mm) Tempered glass side panels and sash
- Glare-free fluorescent lamp mounted outside contamination area
- Blower and light switches are front-mounted for easy access
- Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
- Back panel has two utility ports for service connections
- Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface
For operator safety, UV lamp cannot be turned on when fluorescent lamp is in use.
Order Info
28.75L x 37 in. H (73 x 94cm). 115 V 60 Hz, 3A.
Certifications
ETL listed (certified to UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1).
Compliance
ASHRAE™ 110-1995 conformity tested. Biological containment verified per NSF Standard Number 49 Personnel Protection Test.
Specifications
Specifications
| Product Type | Enclosure |
| Power Consumption | 98 W |
| Height (English) | 37.08 in. |
| Sash Opening (Metric) | 23.97 cm |
| Filter Type | HEPA |
| Housing Material | Epoxy Coated Aluminum and Steel |
| Lighting | Fluorescent |
| Height (Metric) | 94.18 cm |
| Volume | 110 to 155 cfm |
| Width (English) | 23.75 in. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.