Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ HEPA-Filtered Safety Enclosure with Guardian Monitor
SDP

Catalog No. 10304133 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Enclosure
Laminar Flow Safety Cabinet
Electrical Requirements:
115 V, 60 Hz
230 V, 50/60 Hz
Width (Metric):
60.32 cm
90.8 cm
121.28 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Electrical Requirements Width (Metric)
10-304-133 Enclosure 230 V, 50/60 Hz 121.28 cm
10-304-129 Enclosure 230 V, 50/60 Hz 90.8 cm
10-304-131 Laminar Flow Safety Cabinet 115 V, 60 Hz 121.28 cm
10-304-125 Enclosure 230 V, 50/60 Hz 60.32 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 10-304-133 Supplier Labconco™ Supplier No. 3980422
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Labconco Purifier™ HEPA-Filtered Safety Enclosures with Guardian Monitor provide personnel and environmental biohazard protection.

Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.

  • Total exhaust from 155 to 310cfm, depending on hood width
  • Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
  • Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
  • Airfoil with Clean-Sweep™ airflow openings
  • Built-in Guardian™ Airflow Monitor to alert user of face velocities outside preset range (where applicable)
  • Front-mounted pressure gauge monitors pressure differential against the HEPA filter
  • HEPA filter is 99.99% efficient on 0.3μm particles (where applicable)
  • Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
  • Class I Enclosures have a 15 W, 254 nm germicidal UV lamp to use in conjunction with surface disinfection to ensure decontamination of biohazardous particulates while unattended
  • 0.25 in. (6.4mm) Tempered glass side panels and sash
  • Glare-free fluorescent lamp mounted outside contamination area
  • Blower and light switches are front-mounted for easy access
  • Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
  • Back panel has two utility ports for service connections
  • Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface

For operator safety, UV lamp cannot be turned on when fluorescent lamp is in use.

Order Info

28.75L x 37 in. H (73 x 94cm). 115 V 60 Hz, 3A.

Certifications

ETL listed (certified to UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1).

Compliance

ASHRAE™ 110-1995 conformity tested. Biological containment verified per NSF Standard Number 49 Personnel Protection Test.

Specifications

Product Type Enclosure
Power Consumption 156 W
Height (English) 37.08 in.
Sash Opening (Metric) 23.97 cm
Filter Type HEPA
Housing Material Epoxy Coated Aluminum and Steel
Lighting Fluorescent
Height (Metric) 94.18 cm
Volume 220 to 310 cfm
Width (English) 47.75 in.
Width (Metric) 121.28 cm
Frequency 50/60 Hz
Weight (English) 195 lb.
Weight (Metric) 88.45 Kg
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Amperage 1 A
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.