Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, US Models
SDP

Catalog No. 302411101 Shop All Labconco Corporation Products
Change view
Click to view available options
Height (English):
61.7 in.
89.2 to 95.2 in.
Width (English):
3 ft.
4 ft.
5 ft.
6 ft.
Sash Opening (English):
8 in.
10 in.
Includes:
Cabinet
Cabinet, Accessory Package
Cabinet, Accessory Package, Base Stand
Cabinet, Base Stand
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English) Sash Opening (English) Includes
30-241-1101 89.2 to 95.2 in. 4 ft. 10 in. Cabinet, Accessory Package, Base Stand
30-231-0101 89.2 to 95.2 in. 3 ft. 10 in. Cabinet, Base Stand
30-231-1001 61.7 in. 3 ft. 10 in. Cabinet, Accessory Package
30-231-1101 89.2 to 95.2 in. 3 ft. 10 in. Cabinet, Accessory Package, Base Stand
30-238-1101 89.2 to 95.2 in. 3 ft. 8 in. Cabinet, Accessory Package, Base Stand
30-241-1001 61.7 in. 4 ft. 10 in. Cabinet, Accessory Package
30-248-0101 89.2 to 95.2 in. 4 ft. 8 in. Cabinet, Base Stand
30-248-1101 89.2 to 95.2 in. 4 ft. 8 in. Cabinet, Accessory Package, Base Stand
30-251-0101 89.2 to 95.2 in. 5 ft. 10 in. Cabinet, Base Stand
30-251-1001 61.7 in. 5 ft. 10 in. Cabinet, Accessory Package
30-251-1101 89.2 to 95.2 in. 5 ft. 10 in. Cabinet, Accessory Package, Base Stand
30-258-1001 61.7 in. 5 ft. 8 in. Cabinet, Accessory Package
30-261-0001 61.7 in. 6 ft. 10 in. Cabinet
30-261-0101 89.2 to 95.2 in. 6 ft. 10 in. Cabinet, Base Stand
30-261-1001 61.7 in. 6 ft. 10 in. Cabinet, Accessory Package
30-261-1101 89.2 to 95.2 in. 6 ft. 10 in. Cabinet, Accessory Package, Base Stand
30-268-0101 89.2 to 95.2 in. 6 ft. 8 in. Cabinet, Base Stand
30-268-1101 89.2 to 95.2 in. 6 ft. 8 in. Cabinet, Accessory Package, Base Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 30-241-1101 Supplier Labconco™ Supplier No. 302411101
This item is not returnable. View return policy

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models

Specifications

Certifications/Compliance NSF, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel
Width (Metric) Interior 123.2 cm
Product Type Type A2 Biosafety Cabinet
Model Purifier Logic+
Noise Level <63 dBA
Outlets 4
Plug Type North America (115 V, 15 A)
Power Consumption 280 W
Receptacles 2
Height (English) 89.2 to 95.2 in.
Sash Opening (Metric) 25.4 cm
Width (English) Exterior 54.3 in.
Volume (Metric) Exhaust 0.159 to 0.175 m3sec.
Width (Metric) Exterior 137.922 cm
Includes Cabinet, Accessory Package, Base Stand
Lighting LED, UV
Height (Metric) 226.6 to 241.8 cm
Sash Opening (English) 10 in.
Service Fixtures One
Warranty 5 Years
Width (English) Interior 48.5 in.
Width (English) 4 ft.
Width (Metric) 137.922 cm
Frequency 60 Hz
Depth (English) Exterior 31.2 in.
Depth (Metric) Exterior 79.248 cm
Electrical Requirements 115 V, 60 Hz, 12 A
Voltage 115 V
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.