Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purine, 99%

Catalog No. p-3779519
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC131670010

Supplier: Thermo Scientific Chemicals 131670010

Only null left
Add to Cart

CAS: 120-73-0 | C5H4N4 | 120.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-73-0
MDL Number MFCD00079221
InChI Key KDCGOANMDULRCW-UHFFFAOYSA-N
Synonym purine, 9h-purine, 1h-purine, isopurine, beta-purine, 7h-imidazo 4,5-d pyrimidine, imidazo 4,5-d pyrimidine, 3,5,7-triazaindole, 9h-purine van, 3h-purine
PubChem CID 1044
ChEBI CHEBI:17258
IUPAC Name 7H-purine
SMILES C1=C2C(=NC=N1)N=CN2

Specifications

Melting Point 214°C to 217°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C5H4N4
Quantity 1 g
Beilstein 26, 354
Merck Index 15, 8052
Solubility Information Solubility in water: 400g/L water (20°C). Other solubilities: soluble in alcohol,toluene,acetone,hot ethylacet
Molecular Weight (g/mol) 120.11
Formula Weight 120.11
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Purine
Show More Show Less
EINECSNumber 204-421-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.