Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Purine Nucleoside Phosphorylase/PNP Antibody (103), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290053PCP
Description
Purine Nucleoside Phosphorylase/PNP Monoclonal antibody specifically detects Purine Nucleoside Phosphorylase/PNP in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Purine Nucleoside Phosphorylase/PNP | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
103 | |
Western Blot, ELISA, Immunoprecipitation | |
EC 2.4.2.1, FLJ94043, FLJ97288, Inosine phosphorylase, MGC117396, MGC125915, MGC125916, NPFLJ97312, nucleoside phosphorylase, PRO1837, PUNP, purine nucleoside phosphorylase, purine-nucleoside:orthophosphate ribosyltransferase | |
Rabbit immunized with purified, recombinant Human Purine Nucleoside Phosphorylase/PNP (Uniprot#: P00491, Met 1-Ser 289) | |
0.1 mL | |
Neuroscience | |
4860 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction