Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan Cary-Blair Medium

Catalog No. p-7110946
Click to view available options
Fill Volume:
2 mL
5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture.

2 ml Cary-Blair Transport Medium12 x 80 mm screw-cap polypropylene tube, orange cap. Intended for use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture. Premium medical grade plastic components to improve product shelf life. For optimum performance, store at 2-25°C. Made in USA

Specifications

Broth Type Cary Blair Medium
Cap Color Orange
Cap Type Screw
Certifications/Compliance Yes
Color Clear Tube
Content And Storage 2 to 25°C
Description Cary Blair Medium
Format Tubes
Manufacturing Origin United States
Media Type Cary Blair Medium
Sterility Sterile
Fill Volume 2 mL
Packaging Type 50 Bulk
Quantity 50
Dimensions (Dia. x H) 16 x 80 mm
Height (Metric) 80 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.