Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan Lim Broth Medium

Catalog No. 22029650
Change view
Click to view available options
Fill Volume:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
22-029-650 5 mL
22-029-649 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-029-650 Supplier Puritan™ Supplier No. LB500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens as well as in vitro diagnostic use.

  • 5mL Lim Broth with Colistin and Nalidixic Acid.
  • 16 x 100mm screw-cap tube, light blue cap.

Specifications

Broth Type Lim Broth Medium
Cap Color Light Blue
Cap Type Screw
Certifications/Compliance Yes
Color Clear Tube
Content And Storage 2 to 25°C
Description Lim Broth Medium
Format Tubes
Manufacturing Origin United States
Media Type Lim Broth Medium
Sterility Sterile
Fill Volume 5 mL
Incubation Conditions Incubate inoculated MRSA transport medium at 35 ±2°C.
Incubation Time Examine the MRSA transport medium for growth after 18-24 hours.
Packaging Type 50 Bulk
Quantity 50
Dimensions (Dia. x H) 16 x 100 mm
Height (Metric) 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.