Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Dihydrochloride, 10 mg/mL in Distilled Water, Sterile-filtered, Thermo Scientific Chemicals PROMO

Catalog No. J67236XFPM
Click to view available options
:
1 mL

Puromycin (CAS # 58-58-2) is a bacteriostatic antibiotic that acts as an aminoacyl-tRNA analog and disrupts protein synthesis.

General Description

  • Puromycin (puromycine) is an aminoglycoside antibiotic originating from Streptomyces alboniger
  • It acts as an aminoacyl tRNA analog and prematurely terminates peptide elongation
  • Puromycin has effects on both prokaryotes and eukaryotes
  • Puromycin may also have neoplastic activity

Application

  • Puromycin can be used for research use as an immunofluorescent target, in antibiotic resistance testing, or for studies on drug mechanisms
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43 g/mol
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
PubChem CID 439530
ChEBI CHEBI:17939
IUPAC Name (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Type Puromycin Dihydrochloride
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Solubility Information Soluble in Water (50 mg/mL at 20°C).
Chemical Name or Material Puromycin Dihydrochloride
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber AU7350000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.