Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Aminonucleoside, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J66301MCPM
Click to view available options
Quantity:
100 mg

Used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Applications

Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Solubility

Soluble in water at 50 mg/mL. May require heating

Notes

Research Use only: Not intended for animal or human diagnostic or therapeutic use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-60-6
Molecular Formula C12H18N6O3
Molecular Weight (g/mol) 294.32 g/mol
MDL Number MFCD00063462
InChI Key RYSMHWILUNYBFW-GRIPGOBMSA-N
Synonym 3'-Amino-3'-deoxy-N^6,N^6-dimethyladenosine
PubChem CID 6020
ChEBI CHEBI:42839
IUPAC Name (2R,3R,4S,5S)-4-amino-2-[6-(dimethylamino)purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
SMILES CN(C)C1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)N)O

Specifications

Type Puromycin Aminonucleoside
Physical Form Powder
Melting Point 215°C to 216°C
Packaging Plastic Bottle
Quantity 100 mg
Flash Point >110°C (230°F)
Beilstein 93902
Solubility Information Soluble in water. May require heating
Percent Purity 98%
Chemical Name or Material Puromycin aminonucleoside
EINECSNumber 200-388-3
RTECSNumber AU7337000
TSCA Yes
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.