Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Puromycin dihydrochloride
GREENER_CHOICESDP

Catalog No. 408950 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-895-0 Supplier R&D Systems™ Supplier No. 4089/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein synthesis inhibitor

Puromycin dihydrochloride is a protein synthesis inhibitor; leads to the premature release of polypeptide chains as polypeptidyl purine derivatives. Analog of the 3' end of aminoacyl-tRNA. Aminonucleoside antibiotic. Inhibits translation in both in vitro and in vivo systems. Also inhibits the transport of proteins into the mitochondria in vitro.

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 544.43
Product Type Puromycin Dihydrochloride
Purity >98%
Molecular Weight (g/mol) 544.43
Chemical Name or Material Puromycin dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.