Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Purospher™ STAR RP-18 Endcapped (2 μm) Capillary Column, 1 mm I.D.

Catalog No. 01002208 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
01-002-208 150 mm
01-002-206 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-002-208 Supplier MilliporeSigma™ Supplier No. 50115U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ultra-pure silica (99.999%) for excellent peak symmetry.

  • Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, catecholamines, explosives, oligonucleotides, esters, fat soluble vitamins, lipids, fatty acids, flavonoids, ketones, nitrosamines, nucleosides, PAHs, PCBs, peptides, pesticides, phenols, phthalates, preservatives, steroids, metabolized steroids, sulfonamides, sweeteners, and water soluble vitamins.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 1 mm
Length (Metric) 150 mm
Particle Size 2 μm
Pore Size 120 Å
Stationary Phase C18 (octadecyl)
Product Line Purospher STAR
Phase Reversed Phase
Material Stainless Steel
Technique LC/MS
Product Type UHPLC/LC-MS Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.