Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Purvalanol A
GREENER_CHOICESDP

Catalog No. 158010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-801-0 10 mg
15-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-801-0 Supplier Tocris Bioscience™ Supplier No. 1580/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk1, cdk2 and cdk5

Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 4, 4 - 70, 75 - 240 and 100 nM for cdk1, cdk2, cdk5 and cdk 7, respectively). Reversibly arrests synchronised cells in G1 and G2, and inhibits cell proliferation and cell death.

Chemical Identifiers

CAS 212844-53-6
Molecular Formula C19H25ClN6O

Specifications

Quantity 10 mg
Formula Weight 388.9
Purity >98%
Molecular Weight (g/mol) 388.90
Chemical Name or Material Purvalanol A
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.