Learn More
PUS1 Monoclonal Antibody (3A3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524492
Description
Peptide Sequence: MAGNAEPPPA GAACPQDRRS CSGRAGGDRV WEDGEHPAKK LKSGGDEERR EKPPKRKIVL LMAYSGKGYH GMQRNVGSSQ FKTIEDDLVS ALVRSGCIPE NHGEDMRKMS FQRCARTDKG VSAAGQVVSL KVWLIDDILE KINSHLPSHI RILGLKRVTG GFNSKNRCDA RTYCYLLPTF AFAHKDRDVQ DETYRLSAET LQQVNRLLAC YKGTHNFHNF TSQKGPQDPS ACRYILEMYC EEPFVREGLE FAVIRVKGQS FMMHQIRKMV GLVVAIVKGY APESVLERSW GTEKVDVPKA PGLGLVLERV HFEKYNQRFG NDGLHEPLDW AQEEGKVAAF KEEHIYPTII GTERDERSMA QWLSTLPIHN FSATALTAGG TGAKVPSPLE GSEGDGDTD
PUS1 Monoclonal antibody specifically detects PUS1 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
PUS1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9Y606 | |
PUS1 | |
PUS1 (AAH02901, 1 a.a. ∽ 399 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
3A3 | |
Unconjugated | |
PUS1 | |
A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I | |
Mouse | |
Protein A | |
RUO | |
80324 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.