Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566094 Supplier Invitrogen™ Supplier No. PA566094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKVWLIDDILE KINSHLPSHI RILGLKRVTG GFNSKNRCDA RTYCYLLPTF AFAHKDRDVQ DETYRLSAET LQQVNRLLAC YKGTHNFHNF TSQ Highest antigen sequence identity to the following orthologs - mouse 96%, rat 95%.

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PUS1
Gene Accession No. Q9Y606
Gene Alias A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I
Gene Symbols PUS1
Host Species Rabbit
Immunogen Recombinant Human PUS1. Recombinant protein control fragment (Product #RP-105079).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.