Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PUS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568838
Description
This target displays homology in the following species: Mouse: 100%; Rat: 100%.
The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
Specifications
PUS1 | |
Polyclonal | |
Unconjugated | |
PUS1 | |
A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I | |
Rabbit | |
Affinity chromatography | |
RUO | |
56361 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9WU56 | |
PUS1 | |
synthetic peptide corresponding to a region of mouse PUS1 (aa 51-100). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction