Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585285 Supplier Invitrogen™ Supplier No. PA585285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (93%), Sheep (87%). Positive Control: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PUS1
Gene Accession No. Q9WU56, Q9Y606
Gene Alias A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I
Gene Symbols PUS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PUS1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56361, 80324
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.