Learn More
PUS1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301651AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
PUS1 converts uridine into pseudouridine after the nucleotide has been incorporated into RNA. Pseudouridine may have a functional role in tRNAs and may assist in the peptidyl transfer reaction of rRNAs (Chen and Patton, 1999 [PubMed 10094309]).Specifications
| PUS1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9Y606 | |
| PUS1 | |
| Between 377 and 427 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| PUS1 | |
| MLASA1, mitochondrial tRNA pseudouridine synthase A, tRNA pseudouridine synthase A, mitochondrial, tRNA pseudouridine(38-40) synthase, tRNA pseudouridylate synthase I, tRNA uridine isomerase I, tRNA-uridine isomerase I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80324 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.