Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUS1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560908
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0908 Supplier Bethyl Laboratories, Inc Supplier No. A301651A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

PUS1 converts uridine into pseudouridine after the nucleotide has been incorporated into RNA. Pseudouridine may have a functional role in tRNAs and may assist in the peptidyl transfer reaction of rRNAs (Chen and Patton, 1999 [PubMed 10094309]).

Specifications

Antigen PUS1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PUS1
Gene Accession No. Q9Y606
Gene Alias MLASA1, mitochondrial tRNA pseudouridine synthase A, tRNA pseudouridine synthase A, mitochondrial, tRNA pseudouridine(38-40) synthase, tRNA pseudouridylate synthase I, tRNA uridine isomerase I, tRNA-uridine isomerase I
Gene Symbols PUS1
Host Species Rabbit
Immunogen Between 377 and 427
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80324
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.