Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PUS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560808
Description
Immunogen sequence: LVKQEMGKQS LSLGRDDIVQ LKEAYKWITH PLFSEELGVP IDGKSLFEVS VVFAHPETVE DCHFLAAICP DCFKPAKNKQ SVFTRMAVMK A Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Pseudouridylate synthases catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. PUS10 catalyzes the formation of the universal psi55 in the GC loop of transfer RNAs (Probable). Modulator of TRAIL-induced cell death via activation of procaspase 8 and BID cleavage. Required for the progression of the apoptotic signal through intrinsic mitochondrial cell death.
Specifications
PUS10 | |
Polyclonal | |
Unconjugated | |
Pus10 | |
2810013G11Rik; 4933435A13Rik; AU014648; C77560; CCDC139; coiled-coil domain containing 139; coiled-coil domain-containing protein 139; DOBI; pseudouridine synthase 10; pseudouridylate synthase 10; psi55 synthase; Pus10; Putative tRNA pseudouridine synthase Pus10; RGD1306402; tRNA pseudouridine 55 synthase; tRNA pseudouridylate synthase; tRNA-uridine isomerase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150962 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3MIT2 | |
Pus10 | |
Recombinant protein corresponding to Human PUS10. Recombinant protein control fragment (Product #RP-99687). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction