Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUS10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590861 Supplier Invitrogen™ Supplier No. PA590861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAGRYNKYSR NLPQTPWIID GERKLESSVE ELISDHLLAV FKAESFNFSS SGREDVDVRT LGNGRPFAIE LVNPHRVHFT SQEIKELQQK INNSSNKIQV RDLQLVTREA IGHMKEGEEE KTKTYSALIW TNKAIQKKDI EFLNDIKDLK IDQKTPLRVL HRRPLAVRAR VIHFMETQYV DEHHFRLHLK TQAGTYIKEF VHGDFGRTKP NIGSLMNVTA DILELDVESV DVDWPPALDD; Positive Samples: Mouse thymus, Mouse spleen, Mouse testis, Rat thymus.

Pseudouridylate synthases catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. PUS10 catalyzes the formation of the universal psi55 in the GC loop of transfer RNAs (Probable). Modulator of TRAIL-induced cell death via activation of procaspase 8 and BID cleavage. Required for the progression of the apoptotic signal through intrinsic mitochondrial cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUS10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pus10
Gene Accession No. Q9D3U0
Gene Alias 2810013G11Rik; 4933435A13Rik; AU014648; C77560; CCDC139; coiled-coil domain containing 139; coiled-coil domain-containing protein 139; DOBI; pseudouridine synthase 10; pseudouridylate synthase 10; psi55 synthase; Pus10; Putative tRNA pseudouridine synthase Pus10; RGD1306402; tRNA pseudouridine 55 synthase; tRNA pseudouridylate synthase; tRNA-uridine isomerase
Gene Symbols Pus10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 290-529 of human PUS10 (NP_6533102).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305583, 74467
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.