Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUS10 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549267
Description
Pseudouridylate synthases catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. PUS10 catalyzes the formation of the universal psi55 in the GC loop of transfer RNAs (Probable). Modulator of TRAIL-induced cell death via activation of procaspase 8 and BID cleavage. Required for the progression of the apoptotic signal through intrinsic mitochondrial cell death.
Specifications
PUS10 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q3MIT2 | |
PUS10 | |
KLH conjugated synthetic peptide between 52-78 amino acids from the N-terminal region of human PUS10. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PUS10 | |
Putative tRNA pseudouridine synthase Pus10, 5499-, Coiled-coil domain-containing protein 139, tRNA pseudouridine 55 synthase, Psi55 synthase, tRNA pseudouridylate synthase, tRNA-uridine isomerase, PUS10, CCDC139, DOBI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150962 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction