Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUS3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569125
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9125 Supplier Bethyl Laboratories, Inc Supplier No. A304958A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Formation of pseudouridine at position 39 in the anticodon stem and loop of transfer RNAs.

Specifications

Antigen PUS3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PUS3
Gene Accession No. Q9BZE2
Gene Alias 2610020J05Rik, tRNA pseudouridine synthase 3, tRNA pseudouridine(38/39) synthase, tRNA pseudouridylate synthase 3, tRNA-uridine isomerase 3
Gene Symbols PUS3
Host Species Rabbit
Immunogen Between 431 and 481
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83480
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.