Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUS7 Antibody (OTI5F12), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273733FR
Description
PUS7 Monoclonal specifically detects PUS7 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
PUS7 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PUS7 | |
Full length recombinant protein of human PUS7 (NP_061915) produced in HEK293T cells. | |
0.1 mL | |
54517.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI5F12 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 5.4.99.-, FLJ20485, KIAA1897, MGC17720, pseudouridylate synthase 7 homolog, pseudouridylate synthase 7 homolog (S. cerevisiae) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction