Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PUS7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546606 Supplier Invitrogen™ Supplier No. PA546606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FKISEIQLEP NNFPKKPKLD LQNLSLEDGR NQEVHTLIKY TDGDQNHQSG Sequence homology: Human: 100%.

Pseudouridylate synthase that catalyzes pseudouridylation of RNAs. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUS7L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PUS7L
Gene Accession No. Q9H0K6
Gene Alias pseudouridylate synthase 7 homolog (S. cerevisiae)-like; pseudouridylate synthase 7 homolog-like protein; pseudouridylate synthase 7 like; pseudouridylate synthase 7-like; PUS7L
Gene Symbols PUS7L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PUS7L (aa 55-104).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83448
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.