Learn More
PUSL1 Polyclonal Antibody

Description
PUSL1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PUSL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PUSL1 |
| Gene Accession No. | Q8N0Z8 |
| Gene Alias | pseudouridylate synthase-like 1; PUSL1; tRNA pseudouridine synthase-like 1; tRNA pseudouridylate synthase-like 1; tRNA-uridine isomerase-like 1 |
| Gene Symbols | PUSL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.