Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Putative multidrug resistance protein 1 Monoclonal Antibody (1-6)
GREENER_CHOICE

Catalog No. PIMA534893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534893 Supplier Invitrogen™ Supplier No. MA534893
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: Rice.

Multidrug resistance proteins (MRPs) are a subset of the ATP-binding cassette genes that are phase III detoxification proteins involved in transporting exogenous and endogenous compounds across cellular membranes. Energy-dependent efflux pump responsible for decreased drug accumulation in multidrug-resistant cells. The MRPs characterized thus far are organic anion transporters. Many good substrates for these transporters are highly charged and do not penetrate the cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Putative multidrug resistance protein 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1-6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene LOC4326410
Gene Accession No. 0
Gene Alias ABC transporter B family member 11; ABC transporter B family member 11; ABC transporter B family member 9; LOC4326410; mdr6; MDR-like ABC transporter; OJ1029_F04.19; Os01g0533900; OSNPB_010533900; putative multidrug resistance protein 1
Gene Symbols LOC4326410
Host Species Mouse
Immunogen Recombinant protein within rice Putative multidrug resistance protein 1 aa 427-595.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4326410
Target Species Plant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.