Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Puumala Virus Glycoprotein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572799
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572799 Supplier Invitrogen™ Supplier No. PA572799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Puumala Virus Glycoprotein Polyclonal Antibody for ELISA

Puumala Virus Glycoprotein are part of the envelope proteins structure of the Puumala virus (PUUV) which is spread by rodents and is transmitted to humans by inhalation or ingestion of food contaminated with rodent excreta (2). Nephropathia epidemica is endemic in western Russia, Finland, Sweden, France, Belgium, Germany, and former Yugoslavia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Puumala Virus Glycoprotein
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PUUVsMgp1
Gene Symbols PUUVsMgp1
Host Species Rabbit
Immunogen Puumala virus glycoprotein antibody for a 19 amino acid peptide near the amino terminus of the Puumala virus glycoprotein. The immunogen is within amino acids 90 - 140 of Puumala Virus Glycoprotein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2943082
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.