Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puumala Virus Nucleocapsid Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572800 Supplier Thermo Scientific Supplier No. PA572800
Add to Cart
Add to Cart

Puumala Virus Nucleocapsid Polyclonal Antibody

Puumala Virus Nucleocapsid Polyclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Puumala Virus Nucleocapsid
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PUUVsSgp1
Gene Symbols PUUVsSgp1
Host Species Rabbit
Immunogen Puumala virus nucleocapsid antibody for a 19 amino acid peptide near the amino terminus of the Puumala virus nucleocapsid. The immunogen is within amino acids 130 - 180 of Puumala Virus Nucleocapsid.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2943083
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.