Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Mouse anti-Human, Alexa Fluor 647, Clone: TX24 (also known as TX-24), BD

Catalog No. BDB566305 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566305 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566305 Supplier BD Biosciences Supplier No. 566305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TX24 monoclonal antibody specifically binds to the Poliovirus Receptor (PVR) which is also known as CD155, Nectin-like protein 5 (NECL5). PVR is a ñ70 kDa nectin-like type I transmembrane glycoprotein that belongs to the PVR-related (PRR) family within the Ig superfamily. In addition to two cell surface PVR isoforms (alpha and delta), two secreted PVR isoforms (beta and gamma) have been reported that share the same three Ig domains but differ in their C-termini. PVR is expressed on monocytes, macrophages, endothelial cells, epithelia cells, CD34+ thymocytes, and neurons. In addition to serving as a receptor for poliovirus and cytomegalovirus, PVR functions as an adhesion molecule involved in cell-cell and cell-matrix adhesion through interaction with CD96 (TACTILE), Nectin 1-3 (CD111, CD112, CD113), CD226, and vitronectin. PVR promotes natural killer (NK) cell adhesion to and lysis of target cells.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PVR (CD155)
Classification Monoclonal
Clone TX24 (also known as TX-24)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Poliovirus receptor, HVED, Nectin-like protein 5, NECL5, NECL-5, PVS, TAGE4
Host Species Mouse
Immunogen Human PVR Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.