Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Mouse anti-Human, Clone: L95, MilliporeSigma™

Mouse Monoclonal Antibody

$182.31 - $468.00

Specifications

Antigen PVR (CD155)
Clone L95
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays, Neutralization
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF273425
View Documents
MilliporeSigma
MABF273425UG
25 μg
Each for $182.31
Only null left
Add to Cart
 
MABF2734100
View Documents
MilliporeSigma
MABF2734100UG
100 μg
Each for $468.00
Only null left
Add to Cart
 
Description

Description

PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition, Neutralizing.
Specifications

Specifications

PVR (CD155)
1.0 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without azide
IgG1 κ
Protein G purified
L95
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays, Neutralization
Unconjugated
Mouse
Human
Human embryonic 293T cells.
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.