Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Mouse anti-Human, Unconjugated, Clone: SKII.4, MilliporeSigma™

Catalog No. MABF22452MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22452MI 25 μL
MABF22451MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22452MI Supplier MilliporeSigma Supplier No. MABF224525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibits Activity/Function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PVR (CD155)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SKII.4
Conjugate Unconjugated
Gene Accession No. NP_006496.4
Gene Alias Poliovirus receptor;Nectin-like protein 5;NECL-5;CD155
Gene Symbols PVR, PVS
Host Species Mouse
Immunogen SK-N-S1 human neuroblastoma cells
Molecular Weight of Antigen 45.31 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone SKII.4 is a mouse monoclonal antibody that specifically detects human PVR (CD155).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.