Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Rat anti-Mouse, Unconjugated, Clone: TX56, MilliporeSigma™

Catalog No. MABF23491MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23491MI 100 μL
MABF23492MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23491MI Supplier MilliporeSigma Supplier No. MABF2349100UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Inhibits Activity/Function, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PVR (CD155)
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TX56
Conjugate Unconjugated
Gene Alias Poliovirus receptor
Gene Symbols Pvr, cd155, D7ertd458e
Host Species Rat
Immunogen CD155 transfected cells.
Molecular Weight of Antigen 44.66 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone TX56 is a rat monoclonal antibody that specifically detects mouse PVR (CD155).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.