Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PvRBP2b Mouse anti-Plasmodium vivax, Unconjugated, Clone: 3E9, MilliporeSigma™
Mouse Monoclonal Antibody
$204.00 - $516.00
Specifications
| Antigen | PvRBP2b |
|---|---|
| Clone | 3E9 |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF19952MI
|
MilliporeSigma
MABF199525UL |
25 μg |
Each for $204.00
|
|
|||||
|
MABF19951MI
|
MilliporeSigma
MABF1995100UL |
100 μg |
Each for $516.00
|
|
|||||
Description
PvRBP2b Monoclonal antibody specifically detects PvRBP2b in Plasmodium vivax samples. It is validated for ELISA, Flow Cytometry, Inhibits Activity/Function, Western Blotting.Specifications
| PvRBP2b | |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Plasmodium | |
| RBP-2, RBP2 | |
| IgG2a κ | |
| Protein G Purified | |
| Clone 3E9 is a mouse monoclonal antibody that specifically detects Reticulocyte binding protein 2b (PvRBP2b) in Plasmodium vivax. |
| 3E9 | |
| Monoclonal | |
| Purified | |
| Inflammation and Immunology | |
| Reticulocyte-binding protein 2 | |
| His-tagged recombinant fragment corresponding to 1294 amino acids from the N-terminal half of Plasmodium vivax Reticulocyte-binding protein 2 (PvRBP2b). | |
| Primary | |
| Stable for 1 year at -20°C from date of receipt. | |
| ∼120-150 kDa observed; 331.44 kDa calculated (for full length 2pVRBP2b). Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title