Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PvRBP2b Mouse anti-Plasmodium vivax, Unconjugated, Clone: 6H1, MilliporeSigma™

Catalog No. MABF19942MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF19942MI 25 μL
MABF19941MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF19942MI Supplier MilliporeSigma Supplier No. MABF199425UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PvRBP2b Monoclonal antibody specifically detects PvRBP2b in Plasmodium vivax samples. It is validated for ELISA, Flow Cytometry, Inhibits Activity/Function, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PvRBP2b
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6H1
Conjugate Unconjugated
Gene Alias Reticulocyte-binding protein 2
Gene Symbols RBP-2, RBP2
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 1294 amino acids from the N-terminal half of Plasmodium vivax Reticulocyte-binding protein 2 (PvRBP2b).
Molecular Weight of Antigen ∼120-150 kDa observed; 331.44 kDa calculated (for full length 2pVRBP2b). Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 6H1 is a mouse monoclonal antibody that specifically detects Reticulocyte-binding protein 2 (PvRBP2b) in Plasmodium vivax.
Target Species Plasmodium
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.